missing translation for 'onlineSavingsMsg'
Learn More

(Phenylthio)acetic acid, 97%

Product Code. 11445946
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11445946 25 g 25g
11455946 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11445946 Supplier Thermo Scientific Alfa Aesar Supplier No. B22280.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 103-04-8 | C8H7O2S | 167.20 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 103-04-8
Molecular Formula C8H7O2S
Molecular Weight (g/mol) 167.20
MDL Number MFCD00004355
InChI Key MOTOSAGBNXXRRE-UHFFFAOYSA-M
Synonym phenylthio acetic acid, thiophenoxyacetic acid, 2-phenylthio acetic acid, phenylthioglycolic acid, phenylmercaptoacetic acid, 2-phenylsulfanyl acetic acid, phenylsulfanyl acetic acid, acetic acid, phenylthio, phenylmercapto acetic acid, carboxymethyl phenyl sulfide
PubChem CID 59541
IUPAC Name 2-phenylsulfanylacetic acid
SMILES [O-]C(=O)CSC1=CC=CC=C1

Specifications

Melting Point 61°C to 66°C
Odor Stench
Quantity 25 g
Beilstein 2045072
Formula Weight 168.22
Percent Purity 97%
Chemical Name or Material (Phenylthio)acetic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.