missing translation for 'onlineSavingsMsg'
Learn More

Phenylsulfonylacetone, 97%

Product Code. 11468026
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
This item is not returnable. View return policy

Product Code. 11468026

Brand: Thermo Scientific Alfa Aesar A14085.03

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 5000-44-2 | C9H10O3S | 198.236 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5000-44-2
Molecular Formula C9H10O3S
Molecular Weight (g/mol) 198.236
MDL Number MFCD00047794
InChI Key YBLGSNMIIPIRFC-UHFFFAOYSA-N
Synonym phenylsulfonylacetone, benzenesulfonylacetone, 1-phenylsulfonyl propan-2-one, 1-benzenesulfonyl propan-2-one, benzenesulphonylacetone, 1-phenylsulfonyl acetone, phenyl sulfonyl acetone, phenylsulphonylacetone, phenylsulfonyl acetone, 1-phenylsulfonyl-2-propanone
PubChem CID 735827
IUPAC Name 1-(benzenesulfonyl)propan-2-one
SMILES CC(=O)CS(=O)(=O)C1=CC=CC=C1

Specifications

Melting Point 54°C to 58°C
Quantity 1 g
Beilstein 640928
Formula Weight 198.24
Percent Purity 97%
Chemical Name or Material Phenylsulfonylacetone

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.