missing translation for 'onlineSavingsMsg'
Learn More

Phenylsilane, 97%

Product Code. 11409016
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11409016 25 g 25g
11499006 5 g 5g
11419016 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11409016 Supplier Thermo Scientific Alfa Aesar Supplier No. A17901.14
 Restricted Item
Estimated Shipment: 07-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 694-53-1 | C6 H8 Si | 108.22 g/mol

It is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as a pharmaceutical intermediate.

Solubility
Contact with water releases a flammable alcohol. Soluble in water (Contact with water releases flammable gases), and most common organic solvents.

Notes
Moisture Sensitive. Store away from water/moisture. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Protect against electrostatic charges. Keep ignition sources away. Store under inert gas.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 694-53-1
Molecular Formula C6 H8 Si
Molecular Weight (g/mol) 108.22
MDL Number MFCD00013478
InChI Key XJWOWXZSFTXJEX-UHFFFAOYSA-N
Synonym phenylsilane, benzene, silyl, silylbenzene, silane, phenyl, fenylsilan, fenylsilan czech, phenylsilane, silyl, silane, phenyl-,
PubChem CID 6327628
IUPAC Name phenylsilicon
SMILES C1=CC=C(C=C1)[Si]

Specifications

Boiling Point 120°C
Quantity 25 g
UN Number UN3399
Flash Point 7°C (45°F)
Solubility Information Contact with water releases a flammable alcohol. Soluble in water (Contact with water releases flammable gases),and most common organic solvents.
Formula Weight 108.22
Percent Purity 97%
Refractive Index 1.5105
Sensitivity Moisture sensitive
Density 0.88
Chemical Name or Material Phenylsilane
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Phenylsilane
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
  • Substance which, in contact with water, emits flammable gases Category 2
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H261-In contact with water releases flammable gases.
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • EUH014-Reacts violently with water.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P231+P232-Handle under inert gas. Protect from moisture.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
Supplemental information
  • MIXTURE LIST-Contain: Phenylsilane

RUO – Research Use Only

Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.