missing translation for 'onlineSavingsMsg'
Learn More

Phenylselenyl Bromide, TRC

Product Code. 30428246
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
Product Code. Quantity unitSize
30428246 1 g 1g
1 options
This item is not returnable. View return policy

Product Code. 30428246

Brand: Toronto Research Chemicals TRCP3368251G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Phenylselenyl Bromide
CAS 34837-55-3
Molecular Formula C6 H6 Br Se
Quantity 1 g
Synonym Benzeneselenyl Bromide, Phenylselenenyl Bromide, Phenylselenium Bromide
InChI Formula InChI=1S/C6H6BrSe/c7-8-6-4-2-1-3-5-6/h1-5,7H
SMILES Br=[Se]c1ccccc1
IUPAC Name (phenyl-λ3-selanylidene)-λ3-bromane
Molecular Weight (g/mol) 236.976
Formula Weight 236.882
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.