missing translation for 'onlineSavingsMsg'
Learn More

Phenylselenol, 98%, Thermo Scientific Chemicals

Catalog Number p-184161
missing translation for 'orderingAttributeHoverText'
Quantity:
1 g
10 g
Packaging:
Glass bottle
645-96-5
C6H6Se
157
MFCD00000001
645-96-5
C6H6Se
157
MFCD00000001

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color Yellow
Boiling Point 183.0°C
Flash Point 70°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula C6H5SeH
Refractive Index 1.6150 to 1.6180
Quantity 1 g
Formula Weight 157
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material Phenylselenol, 98%
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Phenylselenol
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
  • H301-Toxic if swallowed.
  • H331-Toxic if inhaled.
  • H373-May cause damage to organs.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P273-Avoid release to the environment.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
missing translation for 'delimitedSupplementalStatement'
  • MIXTURE LIST-Contains : Benzeneselenol

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'