missing translation for 'onlineSavingsMsg'
Learn More

Phenylselenenyl bromide, 98%

Product Code. 11433884
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11433884 5 g 5g
11454030 1 g 1g
11443884 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11433884 Supplier Thermo Scientific Alfa Aesar Supplier No. A11906.06
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 34837-55-3 | C6H5BrSe | 235.981 g/mol

Employed in the synthesis of enones by selenenylation of α-cyano ketones with lithium naphthalenide. Phenylselenyl bromide was used in the preparation of exo-2-methyl-7-phenylselenyl-octahydro-8-oxa-2,7b-diaza-dicyclopenta[a,e]pentalene-1,3-dione and endo-2-methyl-7-phenylselanyl-octahydro-8-oxa-2,7b-diaza-dicyclopenta[a,e]pentalene-1,3-dione.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Employed in the synthesis of enones by selenenylation of α-cyano ketones with lithium naphthalenide. Phenylselenyl bromide was used in the preparation of exo-2-methyl-7-phenylselenyl-octahydro-8-oxa-2,7b-diaza-dicyclopenta[a,e]pentalene-1,3-dione and endo-2-methyl-7-phenylselanyl-octahydro-8-oxa-2,7b-diaza-dicyclopenta[a,e]pentalene-1,3-dione.

Solubility
Soluble in toluene, very faint turbidity.

Notes
Moisture Sensitive. Store away from air and water/moisture. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 34837-55-3
Molecular Formula C6H5BrSe
Molecular Weight (g/mol) 235.981
MDL Number MFCD00000047
InChI Key LCEFEIBEOBPPSJ-UHFFFAOYSA-N
Synonym phenylselenyl bromide, benzeneselenenyl bromide, phenylselenenyl bromide, bromoselenobenzene, phsebr, phenylselenylbromide, benzeneselenyl bromide, phenylselanyl bromane, brseph, phenylseleno bromide
PubChem CID 123446
IUPAC Name phenyl selenohypobromite
SMILES C1=CC=C(C=C1)[Se]Br

Specifications

Melting Point 56°C to 62°C
Boiling Point 107°C to 108°C (15 mmHg)
Quantity 5 g
UN Number UN2928
Beilstein 2039772
Sensitivity Moisture sensitive
Solubility Information Soluble in toluene,very faint turbidity.
Formula Weight 235.98
Percent Purity 98%
Chemical Name or Material Phenylselenenyl bromide
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • Phenylselenyl bromide
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Skin corrosion/irritation Category 1 B
  • Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
  • H301-Toxic if swallowed.
  • H314-Causes severe skin burns and eye damage.
  • H331-Toxic if inhaled.
  • H373-May cause damage to organs.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contain: Bromoselenobenzene

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.