missing translation for 'onlineSavingsMsg'
Learn More

Phenylpropiolic acid, 98+%

Product Code. p-7022403
Click to view available options
Quantity:
10 g
25 g
50 g
Unit Size:
10g
25g
50g
This item is not returnable. View return policy

Product Code. 11442158

Brand: Thermo Scientific Alfa Aesar A15163.09

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 637-44-5 | C9H6O2 | 146.15 g/mol

Phenylpropiolic acid is used as a Pharmaceutical Intermediates, laboratory chemicals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Phenylpropiolic acid is used as a Pharmaceutical Intermediates, laboratory chemicals.

Solubility
Soluble in water.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 637-44-5
Molecular Formula C9H6O2
Molecular Weight (g/mol) 146.15
MDL Number MFCD00004361
InChI Key XNERWVPQCYSMLC-UHFFFAOYSA-N
Synonym phenylpropiolic acid, 3-phenylpropiolic acid, phenylpropynoic acid, 3-phenylpropynoic acid, 2-propynoic acid, 3-phenyl, phenylacetylenecarboxylic acid, 3-phenyl-2-propynoic acid, phenylacetylene monocarboxylic acid, propiolic acid, 3-phenyl, phenyl propiolic acid
PubChem CID 69475
IUPAC Name 3-phenylprop-2-ynoic acid
SMILES OC(=O)C#CC1=CC=CC=C1

Specifications

Melting Point 135°C to 138°C
Quantity 10 g
Beilstein 742587
Solubility Information Soluble in water.
Formula Weight 146.15
Percent Purity ≥98%
Chemical Name or Material Phenylpropiolic acid

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.