missing translation for 'onlineSavingsMsg'
Learn More

Phenylpropiolaldehyde diethyl acetal, 98%

Product Code. 11394334
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 11394334

Brand: Thermo Scientific Alfa Aesar L00872.14

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 6142-95-6 | C13H16O2 | 204.27 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6142-95-6
Molecular Formula C13H16O2
Molecular Weight (g/mol) 204.27
MDL Number MFCD00009238
InChI Key DTEGZYXCDQFSBZ-UHFFFAOYSA-N
Synonym 3,3-diethoxyprop-1-yn-1-yl benzene, phenylpropiolaldehyde diethyl acetal, phenylpropargyl aldehyde diethyl acetal, 3-phenylpropynal diethyl acetal, dehydrocinnamaldehyde-diethylacetal, benzene, 3,3-diethoxy-1-propynyl, propiolaldehyde, phenyl-, diethyl acetal, 1,1-diethoxy-3-phenylprop-2-yne, acmc-1bh6o, propiolaldehyde, diethyl acetal
PubChem CID 64818
IUPAC Name 3,3-diethoxyprop-1-ynylbenzene
SMILES CCOC(OCC)C#CC1=CC=CC=C1

Specifications

Density 0.991
Boiling Point 99°C to 100°C (2 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.517
Quantity 25 g
Beilstein 2049715
Formula Weight 204.27
Percent Purity 98%
Chemical Name or Material Phenylpropiolaldehyde diethyl acetal

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.