missing translation for 'onlineSavingsMsg'
Learn More

Phenylphosphonic Dichloride, TRC

Product Code. 30425948
Change view
Click to view available options
Quantity:
10 g
Unit Size:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30425948 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30425948 Supplier Toronto Research Chemicals Supplier No. TRCP33771510G
 Restricted Item
Estimated Shipment: 21-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Phenylphosphonic Dichloride
CAS 824-72-6
Molecular Formula C6H5Cl2OP
Quantity 10 g
Synonym P-Phenylphosphonic dichloride, Benzenephosphonic dichlorideBenzenephosphonyl chloride, Dichlorophenylphosphine oxide, NSC 66477Phenyldichlorophosphine oxide
InChI Formula InChI=1S/C6H5Cl2OP/c7-10(8,9)6-4-2-1-3-5-6/h1-5H
SMILES ClP(=O)(Cl)c1ccccc1
IUPAC Name dichlorophosphorylbenzene
Molecular Weight (g/mol) 194.98
Formula Weight 193.9455
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.