missing translation for 'onlineSavingsMsg'
Learn More

Phenylphosphonic dichloride, 97%

Product Code. 10679833 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
250 mL
1 L
Packaging:
Glass bottle
Unit Size:
1 L
250mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10679833 1 L Glass bottle 1 L
10512151 250 mL Glass bottle 250mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10679833 Supplier Thermo Scientific Acros Supplier No. 130800010
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 824-72-6 | C6H5Cl2OP | 194.98 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 824-72-6
Molecular Formula C6H5Cl2OP
Molecular Weight (g/mol) 194.98
MDL Number MFCD00002070
InChI Key IBDMRHDXAQZJAP-UHFFFAOYSA-N
Synonym phenylphosphonic dichloride, phosphonic dichloride, phenyl, benzenephosphonic dichloride, phenylphosphonyl dichloride, dichlorophenylphosphine oxide, phenyldichlorophosphine oxide, benzenephosphonyl chloride, phenylphosphonic acid dichloride, phenylphosphonodichloridic acid, phosphonic dichloride, p-phenyl
PubChem CID 69990
IUPAC Name dichlorophosphorylbenzene
SMILES ClP(Cl)(=O)C1=CC=CC=C1

Specifications

Melting Point 3.0°C
Color Yellow to Brown
Density 1.3900g/mL
Boiling Point 258.0°C
Flash Point 204°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (Argentometry)
Packaging Glass bottle
Linear Formula C6H5P(O)Cl2
Refractive Index 1.5570 to 1.5610
Quantity 1 L
Beilstein 16, 804
Fieser 01,846; 16,284
Specific Gravity 1.39
Solubility Information Solubility in water: reacts
Formula Weight 194.98
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material Phenylphosphonic dichloride, 97%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.