missing translation for 'onlineSavingsMsg'
Learn More

Phenylphosphonic dichloride, 90+%

Product Code. 11494834
Change view
Click to view available options
Quantity:
50 g
250 g
1000 g
Unit Size:
1000g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11494834 1000 g 1000g
11404844 50 g 50g
11414844 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11494834 Supplier Thermo Scientific Alfa Aesar Supplier No. A13777.0B
 Restricted Item
Estimated Shipment: 07-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 824-72-6 | C6H5Cl2OP | 194.98 g/mol

Phenylphosphonic dichloride is used in the preparation of 2-phenyl-[1,3,2]dioxaphosphepane-2-oxide. Further, it acts as an effective reagent for the chlorinative dehydration of some heterocycles.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Phenylphosphonic dichloride is used in the preparation of 2-phenyl-[1,3,2]dioxaphosphepane-2-oxide. Further, it acts as an effective reagent for the chlorinative dehydration of some heterocycles.

Solubility
Miscible with benzene, chloroform, dimethyl sulfoxide and carbon tetrachloride.

Notes
Moisture sensitive. Store in a cool place. Incompatible with strong oxidizing agents, strong bases and water.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 824-72-6
Molecular Formula C6H5Cl2OP
Molecular Weight (g/mol) 194.98
MDL Number MFCD00002070
InChI Key IBDMRHDXAQZJAP-UHFFFAOYSA-N
Synonym phenylphosphonic dichloride, phosphonic dichloride, phenyl, benzenephosphonic dichloride, phenylphosphonyl dichloride, dichlorophenylphosphine oxide, phenyldichlorophosphine oxide, benzenephosphonyl chloride, phenylphosphonic acid dichloride, phenylphosphonodichloridic acid, phosphonic dichloride, p-phenyl
PubChem CID 69990
IUPAC Name dichlorophosphorylbenzene
SMILES ClP(Cl)(=O)C1=CC=CC=C1

Specifications

Melting Point 1°C to 3°C
Density 1.379
Boiling Point 258°C
Flash Point 204°C (399°F)
Refractive Index 1.56
Quantity 1000 g
UN Number UN3265
Beilstein 1072240
Sensitivity Moisture sensitive
Solubility Information Miscible with benzene,chloroform,dimethyl sulfoxide and carbon tetrachloride.
Formula Weight 194.99
Percent Purity ≥90%
Chemical Name or Material Phenylphosphonic dichloride
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.