missing translation for 'onlineSavingsMsg'
Learn More

Phenylphosphinic acid, 99%, Thermo Scientific™

Product Code. 10704553 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
:
5g; Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
10704553 5g; Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10704553 Supplier Thermo Scientific Acros Supplier No. 130780050
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Specifications

CAS 1779-48-2
Melting Point 82.0°C to 87.0°C
Color White
Boiling Point 180.0°C
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Molecular Formula C6H7O2P
Linear Formula C6H5P(O)(OH)H
MDL Number MFCD00002131
Quantity 5 g
Beilstein 16, 791
Synonym phenylphosphinic acid, benzenephosphinic acid, phosphinic acid, phenyl, benzenephosphonous acid, phenyl phosphinic acid, mlchbqkmvknbov-uhfffaoysa-n, benzolphosphonigs, aure, benzene phosphinic acid, hydroxyphenylphosphineoxide, acmc-1bt5t
Solubility Information Solubility in water: 7.7g/100mL water (25°C). Other solubilities: soluble in alcohol,acetone,slightly soluble in ether,insoluble in benzene,hexane,ccl4
InChI Key MLCHBQKMVKNBOV-UHFFFAOYSA-N
SMILES OP(=O)C1=CC=CC=C1
IUPAC Name hydroxy-oxo-phenylphosphanium
Molecular Weight (g/mol) 142.09
PubChem CID 6327116
Formula Weight 142.09
Percent Purity 99%
Physical Form Crystalline Solid
Chemical Name or Material Phenylphosphinic acid, 99%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Phenylphosphinic acid
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1
  • Specific target organ toxicity Category 3
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.