missing translation for 'onlineSavingsMsg'
Learn More

Phenylmethylsulfonyl Fluoride Crystalline MP Biomedicals

Product Code. 11405362 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11405362 1 g 1g
11415362 5 g 5g
11425362 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11405362 Supplier MP Biomedicals Supplier No. 0219538101
 Restricted Item
Estimated Shipment: 21-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 329-98-6
Molecular Formula C7H7FO2S
Molecular Weight (g/mol) 174.189
InChI Key YBYRMVIVWMBXKQ-UHFFFAOYSA-N
Synonym phenylmethylsulfonyl fluoride, pmsf, benzenemethanesulfonyl fluoride, benzylsulfonyl fluoride, alpha-toluenesulfonyl fluoride, benzylsulphonyl fluoride, alpha-toluenesulphonyl fluoride, phenylmethylsulfonylfluoride, phenylmethylsulphonyl fluoride, .alpha.-toluenesulfonyl fluoride
PubChem CID 4784
ChEBI CHEBI:8102
IUPAC Name phenylmethanesulfonyl fluoride
SMILES C1=CC=C(C=C1)CS(=O)(=O)F

Specifications

Quantity 1 g
Formula Weight 174.2
Physical Form Crystalline
Chemical Name or Material Phenylmethylsulfonyl Fluoride
compliance-icons compliance-icons
Product Identifier
  • Phenylmethylsulfonyl Fluoride
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Skin corrosion/irritation Category 1 B
  • Serious eye damage/eye irritation Category 1
Hazard Statement
  • H301-Toxic if swallowed.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P101-If medical advice is needed, have product container or label at hand.
  • P102-Keep out of reach of children.
  • P103-Read label before use.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P405-Store locked up.
  • P501-Dispose of contents/container to .
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.