missing translation for 'onlineSavingsMsg'
Learn More

Phenylmercuric Acetate, TRC

Product Code. 30426118
Change view
Click to view available options
Quantity:
50 g
Unit Size:
50g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30426118 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30426118 Supplier Toronto Research Chemicals Supplier No. TRCP33563550G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Phenyl-mercury-acetate
CAS 62-38-4
Molecular Formula C8 H8 Hg O2
Quantity 50 g
Synonym Phenylmercuric acetate, (Acetato-κO)phenylmercury, (Acetato)phenylmercury, (Acetato-O)phenylmercury, Phenylmercury Acetate, Acetatophenylmercury, Phenylmercury Deriv. Acetic Acid, Acetoxyphenylmercury, Agrosan, Agrosan D, Algimycin 200, Anticon, Antimucin WBR, Antimucin WDR, (Acetoxymercurio)benzene, Bufen, Bufen 30, Ceresol, Contra Creme, Femma, Fungicide R, Fungitox OR, Hexasan, Hexasan (fungicide), Intercide 60, Intercide PMA 18, Liquiphene, Lorophyn, Meracen, Mercron, Mercuriphenyl acetate, Mercuron, Mergal A 25, Mersolite 8, Mersolite D, NSC 35670, NSC 61321, Neantina, Norforms, Nuodex PMA 18, Nylmerate, PMA, PMA (fungicide), PMA 220, PMAC, PMAS, Panomatic, Parasan, Parasan (bactericide), Phenylmercuric acetate, Phix, Programin, Quicksan, Ruberon, Samtol, Sanitized SPG, Sanmicron, Scutl, Seed Dressing R, Seedtox, Setrete, Spruce Seal, Tag, Tag 331, Tag Fungicide, Tag HL 331, Trigosan, Troysan 30, Troysan PMA 30, Verdasan, Volpar, Zaprawa Nasienna R, Ziarnik
InChI Formula InChI=1S/C6H5.C2H4O2.Hg/c1-2-4-6-5-3-1;1-2(3)4;/h1-5H;1H3,(H,3,4);/q;;+1/p-1
SMILES CC(=O)O[Hg]c1ccccc1
IUPAC Name acetyloxy(phenyl)mercury
Molecular Weight (g/mol) 336.74
Formula Weight 338.0231
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.