missing translation for 'onlineSavingsMsg'
Learn More

Phenylhydrazinium Chloride, Extra Pure, SLR, Fisher Chemical

Product Code. 10141770 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
250 g
Packaging:
Glass Jar
Unit Size:
250g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10141770 250 g Glass Jar 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10141770 Supplier Fisher Chemical Supplier No. P/2600/50

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 59-88-1
Melting Point 250°C
Color White
Packaging Glass Jar
Molecular Formula C6H9ClN2
MDL Number MFCD00012924
Quantity 250 g
Synonym phenylhydrazine hydrochloride, phenylhydrazinium chloride, phenylhydrazine-hcl, phenylhydrazine monohydrochloride, phenylhydrazine hcl, hydrazine, phenyl-, monohydrochloride, unii-h7qfk49svd, phenylhydrazin hydrochlorid, phenylhydrazine.hcl, ccris 5707
InChI Key JOVOSQBPPZZESK-UHFFFAOYSA-N
SMILES [H+].[Cl-].NNC1=CC=CC=C1
IUPAC Name phenylhydrazine;hydrochloride
Molecular Weight (g/mol) 144.60
PubChem CID 60962
Formula Weight 144.61g/mol
Physical Form Powder
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Phenylhydrazine hydrochloride
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • ACUTE AQUATIC Acute 1
  • Carcinogenicity Category 1B
  • Serious eye damage/eye irritation Category 2
  • Germ cell mutagenicity Category 2
  • Respiratory sensitiser Category 1
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 1
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H301-Toxic if swallowed.
  • H372-Causes damage to organs.
  • H400-Very toxic to aquatic life.
  • H350-May cause cancer.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H341-Suspected of causing genetic defects.
  • H311-Toxic in contact with skin.
  • H331-Toxic if inhaled.
Precautionary Statement
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.