missing translation for 'onlineSavingsMsg'
Learn More

Phenylhydrazine hydrochloride, 99%

Product Code. 11407606
Change view
Click to view available options
Quantity:
100 g
Unit Size:
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11407606 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11407606 Supplier Thermo Scientific Alfa Aesar Supplier No. A14645.22
ChemsChrom Mega Deals
Use promo code 27993
View all promotions
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 59-88-1 | C6H9ClN2 | 144.60 g/mol

Phenylhydrazine hydrochloride is used as a selective mannosidase inhibitor. It is used in the study of tyrosine phosphorylation of janus protein tyrosine kinase in the EPO-responsive normal erythroblastoid cells of anemic mice. It is used in steroid assays and as an N-protecting reagent. Its free base, phenylhydrazine is used to prepare indoles, which find application as intermediates in the synthesis of various dyes and pharmaceuticals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Phenylhydrazine hydrochloride is used as a selective mannosidase inhibitor. It is used in the study of tyrosine phosphorylation of janus protein tyrosine kinase in the EPO-responsive normal erythroblastoid cells of anemic mice. It i
TRUSTED_SUSTAINABILITY

Specifications

CAS 59-88-1
Melting Point ∼252°C (decomposition)
Molecular Formula C6H9ClN2
MDL Number MFCD00012924
Quantity 100 g
UN Number UN2811
Beilstein 3594958
Merck Index 14,7293
Synonym phenylhydrazine hydrochloride, phenylhydrazinium chloride, phenylhydrazine-hcl, phenylhydrazine monohydrochloride, phenylhydrazine hcl, hydrazine, phenyl-, monohydrochloride, unii-h7qfk49svd, phenylhydrazin hydrochlorid, phenylhydrazine.hcl, ccris 5707
Solubility Information Soluble in water and alcohol.
InChI Key JOVOSQBPPZZESK-UHFFFAOYSA-N
SMILES [H+].[Cl-].NNC1=CC=CC=C1
IUPAC Name phenylhydrazine;hydrochloride
Molecular Weight (g/mol) 144.60
PubChem CID 60962
Formula Weight 144.61
Percent Purity 99%
Chemical Name or Material Phenylhydrazine hydrochloride
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.