missing translation for 'onlineSavingsMsg'
Learn More

Phenylhydrazine, 97%

Product Code. p-3737206 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Glass bottle
Unit Size:
100g
2.5kg
500g
This item is not returnable. View return policy

Product Code. 10732194

Brand: Thermo Scientific Acros 296681000

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 100-63-0 | C6H8N2 | 108.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 100-63-0
Molecular Formula C6H8N2
Molecular Weight (g/mol) 108.14
MDL Number MFCD00007573
InChI Key HKOOXMFOFWEVGF-UHFFFAOYSA-N
Synonym hydrazinobenzene, hydrazine, phenyl, phenylhydrazin, monophenylhydrazine, phenyl hydrazine, 1-phenylhydrazine, hydrazine-benzene, fenilidrazina, fenylhydrazine, fenylhydrazine dutch
PubChem CID 7516
ChEBI CHEBI:27924
IUPAC Name phenylhydrazine
SMILES NNC1=CC=CC=C1

Specifications

Melting Point 19.0°C
Color Beige or Blue to White
Density 1.0900g/mL
Boiling Point 238.0°C to 241.0°C
Flash Point 89°C
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Linear Formula C6H5NHNH2
Refractive Index 1.6040 to 1.6090
Quantity 100 g
Beilstein 15,II,44
Fieser 01,838; 02,322; 06,457; 07,284
Merck Index 15,7404
Specific Gravity 1.09
Solubility Information Solubility in water: 145g/L (20°C). Other solubilities: soluble in diluted acids,miscible with alcohol,ether,chloroform and,benzene,sparingly soluble in petroleum ether
Formula Weight 108.14
Percent Purity 97%
Physical Form Powder
Chemical Name or Material Phenylhydrazine
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Phenylhydrazine
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • Skin sensitiser Category 1
  • Germ cell mutagenicity Category 2
  • Carcinogenicity Category 1B
  • Specific target organ toxicity Category 1
  • ACUTE AQUATIC Acute 1
Hazard Statement
  • H301-Toxic if swallowed.
  • H311-Toxic in contact with skin.
  • H331-Toxic if inhaled.
  • H315-Causes skin irritation.
  • H317-May cause an allergic skin reaction.
  • H319-Causes serious eye irritation.
  • H341-Suspected of causing genetic defects.
  • H350-May cause cancer.
  • H372-Causes damage to organs.
  • H400-Very toxic to aquatic life.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P311-Call a POISON CENTER/doctor/ .
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P273-Avoid release to the environment.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.