missing translation for 'onlineSavingsMsg'
Learn More

Phenylhydrazine, 97%

Product Code. 11493584
Change view
Click to view available options
Quantity:
50 g
250 g
1000 g
Unit Size:
1000g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11493584 1000 g 1000g
11403594 50 g 50g
11413594 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11493584 Supplier Thermo Scientific Alfa Aesar Supplier No. A11246.0B
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 100-63-0 | C6H8N2 | 108.144 g/mol

Phenyl hydrazine is involved in Fischer indole synthesis to prepare indoles, which find application as intermediates in the pharmaceuticals particularly for the tryptamine drug. In analytical chemistry, it is used to differentiate and separate sugars by forming phenyhydrazones. It is used as an N-protecting reagent and for cleavage of phthaloyl group.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Phenyl hydrazine is involved in Fischer indole synthesis to prepare indoles, which find application as intermediates in the pharmaceuticals particularly for the tryptamine drug. In analytical chemistry, it is used to differentiate and separate sugars by forming phenyhydrazones. It is used as an N-protecting reagent and for cleavage of phthaloyl group.

Solubility
Soluble in dilute acids.

Notes
Air and light sensitive. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 100-63-0
Molecular Formula C6H8N2
Molecular Weight (g/mol) 108.14
MDL Number MFCD00007573
InChI Key HKOOXMFOFWEVGF-UHFFFAOYSA-N
Synonym hydrazinobenzene, hydrazine, phenyl, phenylhydrazin, monophenylhydrazine, phenyl hydrazine, 1-phenylhydrazine, hydrazine-benzene, fenilidrazina, fenylhydrazine, fenylhydrazine dutch
PubChem CID 7516
ChEBI CHEBI:27924
IUPAC Name phenylhydrazine
SMILES NNC1=CC=CC=C1

Specifications

Melting Point 18°C to 20°C
Boiling Point 238°C to 241°C
Flash Point 88°C (190°F)
Assay Percent Range 97%
Quantity 1000 g
UN Number UN2572
Beilstein 606080
Merck Index 14,7293
Solubility Information Soluble in dilute acids
Formula Weight 108.14
Percent Purity 97%
Chemical Name or Material Phenylhydrazine
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Phenylhydrazine
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • ACUTE AQUATIC Acute 1
  • Carcinogenicity Category 1B
  • Serious eye damage/eye irritation Category 2
  • Germ cell mutagenicity Category 2
  • Skin corrosion/irritation Category 2
  • Skin sensitiser Category 1
  • Specific target organ toxicity Category 1
Hazard Statement
  • H301-Toxic if swallowed.
  • H311-Toxic in contact with skin.
  • H315-Causes skin irritation.
  • H317-May cause an allergic skin reaction.
  • H319-Causes serious eye irritation.
  • H331-Toxic if inhaled.
  • H341-Suspected of causing genetic defects.
  • H350-May cause cancer.
  • H372-Causes damage to organs.
  • H400-Very toxic to aquatic life.
Precautionary Statement
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P311-Call a POISON CENTER/doctor/ .
Supplemental information
  • MIXTURE LIST-Contain: Phenylhydrazine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.