missing translation for 'onlineSavingsMsg'
Learn More

Phenylglyoxylic Acid, TRC

Product Code. 30011382
Change view
Click to view available options
Quantity:
2.5 g
5 g
10 g
Unit Size:
10g
2.5g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30011382 2.5 g 2.5g
30007340 10 g 10g
30003629 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30011382 Supplier Toronto Research Chemicals Supplier No. TRCP3275002.5G
 Restricted Item
Estimated Shipment: 25-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 611-73-4
Molecular Formula C8 H6 O3
Molecular Weight (g/mol) 150.131
Synonym Oxophenylacetic Acid, Glycopyrronium Bromide Imp. B (EP), Benzeneacetic acid, α-oxo- (9CI, ACI), Glyoxylic acid, phenyl- (6CI, 7CI, 8CI), α-Oxobenzeneacetic acid (ACI), 2-Oxo-2-phenylacetate, 2-Oxo-2-phenylacetic acid, 2-Oxo-2-phenylethanoic acid, Benzoylformic acid, CAA 0869, Formic acid, benzoyl-, LFC 1849, NSC 28293, Phenylgloxylic acid, Phenylglyoxylic acid, Phenyloxoacetic acid, α-Ketophenylacetic acid
IUPAC Name 2-oxo-2-phenylacetic acid
SMILES OC(=O)C(=O)c1ccccc1

Specifications

Quantity 2.5 g
InChI Formula InChI=1S/C8H6O3/c9-7(8(10)11)6-4-2-1-3-5-6/h1-5H,(H,10,11)
Formula Weight 150.032
Percent Purity >95%
Chemical Name or Material Phenylglyoxylic Acid
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.