missing translation for 'onlineSavingsMsg'
Learn More

Phenylglyoxal monohydrate, 97%

Product Code. 10674832 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10674832 5 g Glass bottle 5g
10700861 25 g Glass bottle 25g
10762115 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10674832 Supplier Thermo Scientific Acros Supplier No. 161970050
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1075-06-5 | C8H6O2·H2O | 152.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1075-06-5
Molecular Formula C8H6O2·H2O
Molecular Weight (g/mol) 152.15
MDL Number MFCD00149499
InChI Key NBIBDIKAOBCFJN-UHFFFAOYSA-N
Synonym dihydroxyacetophenone, acetophenone, dihydroxy, dihydroxy 1-phenylethanone, 2,2-dihydroxy-1-phenyl-ethanone, ethanone, 2,2-dihydroxy-1-phenyl, acetophenone, 2,2-dihydroxy, alpha,alpha-dihydroxyacetophenone, dioxyacetophenone, alpha,alpha-dihydroxy-acetophenone
PubChem CID 99611
IUPAC Name 2,2-dihydroxy-1-phenylethanone
SMILES C1=CC=C(C=C1)C(=O)C(O)O

Specifications

Melting Point 71.0°C to 76.0°C
Color Pink or White to Yellow
Boiling Point 142.0°C (125.0 mmHg)
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Linear Formula C6H5COCHO·H2O
Quantity 5 g
Beilstein 07, 670
Formula Weight 152.15
Percent Purity 97%
Physical Form Powder
Chemical Name or Material Phenylglyoxal monohydrate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.