missing translation for 'onlineSavingsMsg'
Learn More

Phenylglyoxal monohydrate, 97%

Product Code. 11463434
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11463434 100 g 100g
11443434 5 g 5g
11453434 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11463434 Supplier Thermo Scientific Alfa Aesar Supplier No. A10677.22
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1075-06-5 | C8H8O3 | 152.149 g/mol

Phenylglyoxal monohydrate was used to modify pig muscle carbonic anhydraseIII and as a specific reagent for arginine groups. It was also used to prepare pyrrolinone and furan derivatives and as as a chemiluminescent reagent for the determination of purines.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Phenylglyoxal monohydrate was used to modify pig muscle carbonic anhydraseIII and as a specific reagent for arginine groups. It was also used to prepare pyrrolinone and furan derivatives and as as a chemiluminescent reagent for the determination of purines.

Solubility
Partly miscible with water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1075-06-5
Molecular Formula C8H8O3
Molecular Weight (g/mol) 152.149
MDL Number MFCD00149499
InChI Key NBIBDIKAOBCFJN-UHFFFAOYSA-N
Synonym dihydroxyacetophenone, acetophenone, dihydroxy, dihydroxy 1-phenylethanone, 2,2-dihydroxy-1-phenyl-ethanone, ethanone, 2,2-dihydroxy-1-phenyl, acetophenone, 2,2-dihydroxy, alpha,alpha-dihydroxyacetophenone, dioxyacetophenone, alpha,alpha-dihydroxy-acetophenone
PubChem CID 99611
IUPAC Name 2,2-dihydroxy-1-phenylethanone
SMILES C1=CC=C(C=C1)C(=O)C(O)O

Specifications

Melting Point 71°C to 77°C
Boiling Point 142°C (125 mmHg)
Quantity 100 g
Beilstein 1854721
Solubility Information Partly miscible with water.
Formula Weight 152.15 (134.12 Anhydrous)
Percent Purity 97%
Chemical Name or Material Phenylglyoxal monohydrate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.