missing translation for 'onlineSavingsMsg'
Learn More

Phenylethyl 3,4-dihydroxycinnamate, 98+%

Product Code. 15493989
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15493989 250 mg 250mg
15483989 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15493989 Supplier Thermo Scientific Alfa Aesar Supplier No. J61386.MD
 Restricted Item
Estimated Shipment: 09-09-2026
Add to basket
This item is not returnable. View return policy

Inhibitor of protein tyrosine kinase, and is cytotoxic to cancer cell lines | CAS: 104594-70-9 | C17H16O4 | 284.31 g/mol

Phenylethyl 3,4-dihydroxycinnamate is a potent and specific inhibitor of nuclear transcription factor, NF-κB, activation. It has been shown to significantly suppress the lipoxygenase pathway of arachidonic acid metabolism during inflammation. It inhibits HIV-1 integrase and also inhibits proliferation of transformed cells. It induces apoptosis in transformed fibroblasts, and interferes with EGF signal transduction affecting both protein kinase C and ornithine decarboxylase activity.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Phenylethyl 3,4-dihydroxycinnamate is a potent and specific inhibitor of nuclear transcription factor, NF-κB, activation. It has been shown to significantly suppress the lipoxygenase pathway of arachidonic acid metabolism during inflammation. It inhibits HIV-1 integrase and also inhibits proliferation of transformed cells. It induces apoptosis in transformed fibroblasts, and interferes with EGF signal transduction affecting both protein kinase C and ornithine decarboxylase activity.

Solubility
Soluble in ethyl acetate at 50mg/ml. Soluble in DMSO and ethanol.

Notes
Light sensitive. Incompatible with oxidizing agents and heat.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 104594-70-9
Molecular Formula C17H16O4
Molecular Weight (g/mol) 284.31
MDL Number MFCD00866470
InChI Key SWUARLUWKZWEBQ-VQHVLOKHSA-N
Synonym caffeic acid phenethyl ester, phenethyl caffeate, cape, capeee, phenethyl 3-3,4-dihydroxyphenyl acrylate, phenylethyl caffeate, unii-g960r9s5sk, caffeic acid phenylethyl ester, 2-phenylethyl caffeate, caffeic acid 2-phenylethyl ester
PubChem CID 5281787
ChEBI CHEBI:8062
IUPAC Name 2-phenylethyl (E)-3-(3,4-dihydroxyphenyl)prop-2-enoate
SMILES OC1=CC=C(\C=C\C(=O)OCCC2=CC=CC=C2)C=C1O

Specifications

Melting Point 127°C to 129°C
Color White to Yellow
Quantity 250 mg
Sensitivity Light sensitive
Solubility Information Soluble in ethyl acetate at 50mg/ml; Soluble in DMSO and ethanol.
Formula Weight 284.31
Percent Purity ≥99%
Physical Form Crystalline Powder
Chemical Name or Material Phenylethyl 3,4-dihydroxycinnamate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.