missing translation for 'onlineSavingsMsg'
Learn More

Phenylephrine hydrochloride, MedChemExpress

Product Code. p-200096875
Encompass_Preferred
Click to view available options
Quantity:
100 mg
500 mg
Unit Size:
100mg
500mg
This item is not returnable. View return policy

Product Code. 16488326

Brand: Medchem Express HYB0471100mg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress (R)-(-)-Phenylephrine hydrochloride is a selective α1-adrenoceptor agonist with pKis of 5.86, 4.87 and 4.70 for α1D, α1B and α1A receptors respectively.

(R)-(-)-Phenylephrine hydrochloride is a selective α1-adrenoceptor agonist with pKis of 5.86, 4.87 and 4.70 for α1D, α1B and α1A receptors respectively.

Specifications

Chemical Name or Material Phenylephrine hydrochloride
CAS 61-76-7
Color White
Purity Grade Notes Research
Molecular Formula C9H14ClNO2
Quantity 100 mg
Synonym (R)-(-)-Phenylephrine hydrochlorideL-Phenylephrine hydrochloride
Solubility Information DMSO : ≥ 150 mg/mL (736.49 mM) ∣H2O : 100 mg/mL (490.99 mM; Need ultrasonic)
SMILES OC1=CC([C@@H](O)CNC)=CC=C1.Cl
Molecular Weight (g/mol) 203.67
Formula Weight 203.67
Percent Purity 99.76%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.