missing translation for 'onlineSavingsMsg'
Learn More

Phenylephrine hydrochloride, MedChemExpress

Product Code. 16488326
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
500 mg
Unit Size:
100mg
500mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16488326 100 mg 100mg
16482146 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16488326 Supplier Medchem Express Supplier No. HYB0471100mg
 Restricted Item
Estimated Shipment: 03-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress (R)-(-)-Phenylephrine hydrochloride is a selective α1-adrenoceptor agonist with pKis of 5.86, 4.87 and 4.70 for α1D, α1B and α1A receptors respectively.

(R)-(-)-Phenylephrine hydrochloride is a selective α1-adrenoceptor agonist with pKis of 5.86, 4.87 and 4.70 for α1D, α1B and α1A receptors respectively.

Specifications

Chemical Name or Material Phenylephrine hydrochloride
CAS 61-76-7
Color White
Purity Grade Notes Research
Molecular Formula C9H14ClNO2
Quantity 100 mg
Synonym (R)-(-)-Phenylephrine hydrochlorideL-Phenylephrine hydrochloride
Solubility Information DMSO : ≥ 150 mg/mL (736.49 mM) ∣H2O : 100 mg/mL (490.99 mM; Need ultrasonic)
SMILES OC1=CC([C@@H](O)CNC)=CC=C1.Cl
Molecular Weight (g/mol) 203.67
Formula Weight 203.67
Percent Purity 99.76%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.