missing translation for 'onlineSavingsMsg'
Learn More

Phenylbutazone, TRC

Product Code. 30424922
Change view
Click to view available options
Quantity:
25 g
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30424922 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30424922 Supplier Toronto Research Chemicals Supplier No. TRCP31957025G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Phenylbutazone
CAS 50-33-9
Purity Grade Notes HPLC
Molecular Formula C19 H20 N2 O2
Quantity 25 g
Synonym 3,5-Pyrazolidinedione','4-butyl-1,2-diphenyl-, 1,2-Diphenyl-3,5-dioxo-4-butylpyrazolidine, 1,2-Diphenyl-3,5-dioxo-4-n-butylpyrazolidine, 1,2-Diphenyl-4-butyl-3,5-pyrazolidinedione, 3,5-Dioxo-1,2-diphenyl-4-n-butylpyrazolidine, 4-Butyl-1,2-diphenyl-3,5-dioxopyrazolidine, 4-Butyl-1,2-diphenylpyrazolidine-3,5-dione, 4-n-Butyl-1,2-diphenyl-3,5-pyrazolidinedione, Alindor, Alkazone, Antadol, Anuspiramin, Arthrizon, Artrizin, Artropan, Azolid, Benzone, Betazed, Bizolin 200, Bunetzone, Butacote, Butadion, Butadione, Butalidon, Butapirazol, Butapirazole, Butartrina, Butatron, Butazina, Butazolidin, Butazolidine, Butidiona, Butoz, Buzon, Diphebuzol, Diphenylbutazone, Ecobutazone, Equipalazone, Fenibutal, Fenibutazona, Fenilbutina, Fenotone, Fenylbutazon, Flexazone, G 13,871, Ia-But, Kadol, Mephabutazon, NSC 25134, Phebuzine, Phen-Buta, Phenylbutazon, Phenylbutazone, Pirarreumol B, Pyrabutol, R-3-ZON, Reudo, Reudox, Reumazin, Reumuzol, Robizone, Shigrodin, Tencodyne, Tevcodyne, Ticinil, Todalgil, Uzone, VAC-10, Zolaphen
InChI Formula InChI=1S/C19H20N2O2/c1-2-3-14-17-18(22)20(15-10-6-4-7-11-15)21(19(17)23)16-12-8-5-9-13-16/h4-13,17H,2-3,14H2,1H3
SMILES CCCCC1C(=O)N(N(C1=O)c2ccccc2)c3ccccc3
IUPAC Name 4-butyl-1,2-diphenylpyrazolidine-3,5-dione
Molecular Weight (g/mol) 308.37
Formula Weight 308.1525
Percent Purity >95
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.