missing translation for 'onlineSavingsMsg'
Learn More

Phenylboronic acid neopentylglycol ester, 97%

Product Code. 11936151 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11936151 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11936151 Supplier Thermo Scientific Acros Supplier No. 445500050

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 5123-13-7 | C11H15BO2 | 190.05 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 5123-13-7
Melting Point 65°C to 67°C
Assay Percent Range 96% min. (GC)
Molecular Formula C11H15BO2
MDL Number MFCD05663851
Quantity 5 g
Synonym 5,5-dimethyl-2-phenyl-1,3,2-dioxaborinane, 5,5-dimethyl-1,3,2-dioxaborinan-2-yl benzene, phenylboronic acid neopentyl glycol ester, 1,3,2-dioxaborinane, 5,5-dimethyl-2-phenyl, amtb968, fqmsnyzdfmwlgc-uhfffaoysa-n, phenylboronic acid,neopentyl ester, phenylboronic acid neopentylglycol ester, phenyl boronic acid neopentylglycol ester
InChI Key FQMSNYZDFMWLGC-UHFFFAOYSA-N
SMILES CC1(C)COB(OC1)C1=CC=CC=C1
IUPAC Name 5,5-dimethyl-2-phenyl-1,3,2-dioxaborinane
Molecular Weight (g/mol) 190.05
PubChem CID 11788741
Formula Weight 190.05
Percent Purity 97%
Chemical Name or Material Phenylboronic acid neopentylglycol ester
Show More Show Less
compliance-icons
Product Identifier
  • Phenylboronic acid neopentyl glycol ester
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.