missing translation for 'onlineSavingsMsg'
Learn More

Phenylboronic acid, 98+%, may contain varying amounts of anhydride, ACROS Organics™

Brand:  Acros Organics 130360100

Code : 29

Additional Details : CAS Number : 98-80-6 Weight : 0.04512kg


Packaging Glass bottle
Quantity 10g

 View more versions of this product

Product Code. 10041420

  • £21.30 / 10g

In stock, ready to ship: 2

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Chemical Identifiers

98-80-6
121.93
benzeneboronic acid, boronic acid, phenyl, phenyl boronic acid, phenylboric acid, dihydroxyphenylborane, phenyldihydroxyborane, dihydroxy phenyl borane, phenylboronicacid, borophenylic acid, usaf bo-2
CHEBI:44923
B(C1=CC=CC=C1)(O)O
C6H7BO2
HXITXNWTGFUOAU-UHFFFAOYSA-N
66827
phenylboronic acid

Specifications

Phenylboronic acid
Glass bottle
217°C to 222°C
98-80-6
100.0
C6H7BO2
16, 920
15, 1070
Solubility in water: 10g/L (20°C). Other solubilities: solubility in benzene : 1.75%, xylene : 1.2%, methanol : 178%, ether : 30.2%
B(C1=CC=CC=C1)(O)O
121.93
CHEBI:44923
Crystalline Powder
Authentic
White
10g
96.5
98% min. (HPLC)
C6H5B(OH)2
01,833; 02,317; 03,221; 05,513; 07,284; 09,23; 11,408; 15,254
benzeneboronic acid, boronic acid, phenyl, phenyl boronic acid, phenylboric acid, dihydroxyphenylborane, phenyldihydroxyborane, dihydroxy phenyl borane, phenylboronicacid, borophenylic acid, usaf bo-2
HXITXNWTGFUOAU-UHFFFAOYSA-N
phenylboronic acid
66827
121.93
98+%
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Phenylboronic acid

Signal Word
  • Warning

Hazard Category
  • Acute toxicity Category 4

Hazard Statement
  • H302-Harmful if swallowed.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

Supplemental information
  • MIXTURE LIST-Contains : Boronic acid, phenyl-

SDS Documents