missing translation for 'onlineSavingsMsg'
Learn More

Phenylboronic acid, 98+%, may contain varying amounts of anhydride, Thermo Scientific Chemicals

Product Code. 10440194
Click to view available options
Quantity:
10 g
50 g
250 g
Packaging:
Glass bottle

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 98-80-6
Molecular Formula C6H7BO2
Molecular Weight (g/mol) 121.93
MDL Number MFCD00002103
InChI Key HXITXNWTGFUOAU-UHFFFAOYSA-N
Synonym benzeneboronic acid, boronic acid, phenyl, phenyl boronic acid, phenylboric acid, dihydroxyphenylborane, phenyldihydroxyborane, dihydroxy phenyl borane, phenylboronicacid, borophenylic acid, usaf bo-2
PubChem CID 66827
ChEBI CHEBI:44923
IUPAC Name phenylboronic acid
SMILES OB(O)C1=CC=CC=C1

Specifications

CAS Min % 96.5
CAS Max % 100.0
Melting Point 217°C to 222°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 98% min. (HPLC)
Packaging Glass bottle
Linear Formula C6H5B(OH)2
Quantity 250 g
Beilstein 16, 920
Fieser 01,833; 02,317; 03,221; 05,513; 07,284; 09,23; 11,408; 15,254
Merck Index 15, 1070
Solubility Information Solubility in water: 10g/L (20°C). Other solubilities: solubility in benzene : 1.75%, xylene : 1.2%, methanol : 178%, ether : 30.2%
Formula Weight 121.93
Percent Purity 98+%
Physical Form Crystalline Powder
Chemical Name or Material Phenylboronic acid
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'