missing translation for 'onlineSavingsMsg'
Learn More

Phenylarsonic acid, 97%

Product Code. 11463334
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11463334 25 g 25g
11453334 5 g 5g
11473334 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XVII to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.
Product Code. 11463334 Supplier Thermo Scientific Alfa Aesar Supplier No. A10625.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XVII to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.

CAS: 98-05-5 | C6H7AsO3 | 202.041 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 98-05-5
Molecular Formula C6H7AsO3
Molecular Weight (g/mol) 202.041
MDL Number MFCD00002097
InChI Key LVKZSFMYNWRPJX-UHFFFAOYSA-N
Synonym benzenearsonic acid, monophenylarsonic acid, arsonic acid, phenyl, unii-57f9ku116m, kyselina benzenarsonova czech, arsonic acid, as-phenyl, kyselina benzenarsonova, benzenarsonic acid, benzenearsonie acid, acmc-20ajwq
PubChem CID 7365
ChEBI CHEBI:29851
IUPAC Name phenylarsonic acid
SMILES C1=CC=C(C=C1)[As](=O)(O)O

Specifications

Melting Point 158°C to 162°C (decomposition)
Quantity 25 g
UN Number UN3465
Merck Index 14,1067
Formula Weight 202.04
Percent Purity 97%
Chemical Name or Material Phenylarsonic acid
compliance-icons compliance-icons
Product Identifier
  • Phenylarsonic acid
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
Hazard Statement
  • H301-Toxic if swallowed.
  • H331-Toxic if inhaled.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P273-Avoid release to the environment.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
Supplemental information
  • MIXTURE LIST-Contain: Phenylarsonic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.