missing translation for 'onlineSavingsMsg'
Learn More

Phenylarsine oxide, 97%

Product Code. 11417656
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11417656 5 g 5g
11407656 1 g 1g
11427656 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XVII to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.
Product Code. 11417656 Supplier Thermo Scientific Alfa Aesar Supplier No. A14674.06
 Restricted Item
Estimated Shipment: 05-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XVII to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.

CAS: 637-03-6 | C6H5AsO | 168.03 g/mol

Phenylarsine oxide is used as a poultry coccidiostat in veterinary medicine. It is also used in a membrane-permeable protein tyrosine phosphatase inhibitor.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Phenylarsine oxide is used as a poultry coccidiostat in veterinary medicine. It is also used in a membrane-permeable protein tyrosine phosphatase inhibitor.

Solubility
Soluble in dimethyl sulfoxide, chloroform, benzene, ether, ethanol, 50:50 dimethyl sulfoxide:1 M hydrochloric acid (3.0 mg/ml) and toluene. Insoluble in water.

Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 637-03-6
Molecular Formula C6H5AsO
Molecular Weight (g/mol) 168.03
MDL Number MFCD00001990
InChI Key BQVCCPGCDUSGOE-UHFFFAOYSA-N
Synonym phenylarsine oxide, oxophenylarsine, arzene, phenylarsenoxide, arsenosobenzene, arsine, oxophenyl, phenyl arsine oxide, benzene, arsenoso, fenylarsinoxid, caswell no. 060
PubChem CID 4778
ChEBI CHEBI:75253
IUPAC Name arsorosobenzene
SMILES O=[As]C1=CC=CC=C1

Specifications

Melting Point 140°C to 160°C
Quantity 5 g
UN Number UN3465
Beilstein 2935227
Merck Index 14,6949
Solubility Information Soluble in dimethyl sulfoxide,chloroform,benzene,ether,ethanol,50:50 dimethyl sulfoxide:1 M hydrochloric acid (3.0mg/ml) and toluene. Insoluble in water.
Formula Weight 168.03
Percent Purity 97%
Chemical Name or Material Phenylarsine oxide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.