missing translation for 'onlineSavingsMsg'
Learn More

Phenylalanylglycine, TRC

Product Code. 30422935
Change view
Click to view available options
Quantity:
50 mg
100 mg
500 mg
Unit Size:
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30422935 500 mg 500mg
30426540 50 mg -
30427971 100 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30422935 Supplier Toronto Research Chemicals Supplier No. TRCP399360500MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 721-90-4
Molecular Formula C11H14N2O3
Molecular Weight (g/mol) 222.24
Synonym L-Phenylalanylglycine, 105: PN: US20130123467 SEQID: 128 Claimed Protein, 10: PN: WO2014063098 TABLE: 5 Claimed Protein, 2-[(2S)-2-Amino-3-phenylpropanamido]acetic Acid, 2-[[(2S)-2-Amino-3-phenylpropanoyl]amino]acetic Acid, 28: PN: WO2016049036 SEQID: 69 Claimed Protein, 3: PN: RU2506269 PAGE: 14 Claimed Sequence, NSC 89182
IUPAC Name 2-[[(2S)-2-amino-3-phenylpropanoyl]amino]acetic acid
SMILES N[C@@H](Cc1ccccc1)C(=O)NCC(=O)O

Specifications

InChI Formula InChI=1S/C11H14N2O3/c12-9(11(16)13-7-10(14)15)6-8-4-2-1-3-5-8/h1-5,9H,6-7,12H2,(H,13,16)(H,14,15)/t9-/m0/s1
Quantity 500 mg
Formula Weight 222.1004
Shipping Condition Room Temperature
Chemical Name or Material Phenylalanylglycine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.