missing translation for 'onlineSavingsMsg'
Learn More

Phenylalanine (L-Phenylalanine), LoGiCal

Product Code. 30458390
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30458390 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30458390 Supplier LoGiCal Supplier No. LGCFOR1560.00

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

LoGiCal delivers certified reference materials you can trust as neats, native solutions and stable isotope labelled solutions, with precise documentation and expert support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.

Phenylalanine (L-Phenylalanine)
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material L-Phenylalanine
CAS 63-91-2
Analyte or Component Names L-Phenylalanine
Shipping Condition Room Temperature
Molecular Formula C9 H11 N O2
Certifications/Compliance ISO 9001
Synonym Nateglinide Imp. D (EP), Tyrosine Imp. A (EP), L-Phenylalanine, Alanine, phenyl-, L- (4 CI,7 CI,8 CI), (-)-Phenylalanine, GV 112054 X, GV 112054 X, GV 112054 X, GV 112054 X, GV 112054 X, GV 112054 X, GV 112054 X, GV 112054 X, GV 112054 X, GV 112054 X, GV 112054 X, GV 112054 X, GV 112054 X, GV 112054 X, GV 112054 X, GV 112054 X, GV 112054 X, GV 112054 X, GV 112054 X, GV 112054 X, GV 112054 X, GV 112054 X, GV 112054 X, GV 112054 X, GV 112054 X, GV 112054 X, GV 112054 X, GV 112054 X, GV 112054 X, GV 112054 X, GV 112054 X
InChI Formula InChI=1 S/C9H11NO2/c10-8(9(11)12)6-7-4-2-1-3-5-7/h1-5,8 H,6,10H2,(H,11,12)/t8-/m0/s1
SMILES N[C@@H](Cc1ccccc1)C(=O)O
IUPAC Name (2 S)-2-amino-3-phenylpropanoic acid
Industry Type Forensics
Molecular Weight (g/mol) 165.19
Quantity 10 mg
Formula Weight 165.079 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.