missing translation for 'onlineSavingsMsg'
Learn More

Phenylacetyl Disulfide, TRC

Product Code. 30412403
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30412403 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30412403 Supplier Toronto Research Chemicals Supplier No. TRCP335598100MG
 Restricted Item
Estimated Shipment: 26-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Phenylacetyl Disulfide
CAS 15088-78-5
Molecular Formula C16 H14 O2 S2
Quantity 100 mg
Synonym Bis(phenylacetyl) Disulfide, Di(phenylacetyl) Disulfide, PADS
InChI Formula InChI=1S/C16H14O2S2/c17-15(11-13-7-3-1-4-8-13)19-20-16(18)12-14-9-5-2-6-10-14/h1-10H,11-12H2
SMILES O=C(Cc1ccccc1)SSC(=O)Cc2ccccc2
IUPAC Name S-(2-phenylacetyl)sulfanyl 2-phenylethanethioate
Molecular Weight (g/mol) 302.411
Formula Weight 302.044
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.