missing translation for 'onlineSavingsMsg'
Learn More

Phenylacetic Acid, Mikromol™

Product Code. 17974546
Change view
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17974546 100 mg 100mg
17954596 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: This product contains a category 2 drug precursor according to Regulation (EC) No 273/2004, all customers must complete a declaration before delivery.
Product Code. 17974546 Supplier Mikromol Supplier No. MM0160.03
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Regulatory: This product contains a category 2 drug precursor according to Regulation (EC) No 273/2004, all customers must complete a declaration before delivery.

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Phenylacetic Acid
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Phenylacetic Acid
CAS 103-82-2
Analyte or Component Names Phenylacetic Acid
Shipping Condition Room Temperature
Molecular Formula C8 H8 O2
Certifications/Compliance ISO 17025
Synonym Benzylpenicillin Benzathine Imp. B (EP), Benzylpenicillin Potassium Imp. B (EP), Benzylpenicillin Procaine Imp. E (EP), Cyclopentolate Hydrochloride Imp. B (EP), Cyclopentolate Imp. B (EP), Tropicamide Imp. D (EP), Tropicamide USP Related Compound D, Tropicamide USP RC D, Phenylacetic Acid, Benzeneacetic acid, Acetic acid, phenyl- (6CI,8CI), 2-Phenylacetic acid, NSC 125718, PAA, Phenylethanoic acid, alpha-Toluic acid, omega-Phenylacetic acid, BRL-2853, Benzylpenicillin Potassium Ph Eur Impurity B, BRL-955, USP Amoxicillin Related Compound A, Ph Eur Amoxicillin Impurity A, Penicinic acid (1), Penicinic acid (2), 8-Hydroxypenillic acid (8-HPA), Phenylacetic acid, Tropicamide Related Compound D, Benzylpenicillin Benzathine Impurity B, Benzylpenicillin Potassium Impurity B, Benzylpenicillin Procaine Impurity E, Carbenicillin Sodium Impurity B, Cyclopentolate Hydrochloride Impurity B, Tropicamide Impurity D, Carbenicillin Impurity B, Cyclopentolate Impurity B, Benzylpenicillin Impurity B
InChI Formula InChI=1S/C8H8O2/c9-8(10)6-7-4-2-1-3-5-7/h1-5H,6H2,(H,9,10)
SMILES OC(=O)Cc1ccccc1
IUPAC Name 2-phenylacetic acid
Molecular Weight (g/mol) 136.15
Quantity 100 mg
Formula Weight 136.0524 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.