missing translation for 'onlineSavingsMsg'
Learn More

Phenylacetic Acid, Mikromol™

Product Code. 17954596
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
This item is not returnable. View return policy
Regulatory: This product contains a category 2 drug precursor according to Regulation (EC) No 273/2004, all customers must complete a declaration before delivery.

Product Code. 17954596

Brand: Mikromol MM0160.030025

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: This product contains a category 2 drug precursor according to Regulation (EC) No 273/2004, all customers must complete a declaration before delivery.

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Phenylacetic Acid
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Phenylacetic Acid
CAS 103-82-2
Analyte or Component Names Phenylacetic Acid
Shipping Condition Room Temperature
Molecular Formula C8 H8 O2
Certifications/Compliance ISO 17025
Synonym Benzylpenicillin Benzathine Imp. B (EP), Benzylpenicillin Potassium Imp. B (EP), Benzylpenicillin Procaine Imp. E (EP), Cyclopentolate Hydrochloride Imp. B (EP), Cyclopentolate Imp. B (EP), Tropicamide Imp. D (EP), Tropicamide USP Related Compound D, Tropicamide USP RC D, Phenylacetic Acid, Benzeneacetic acid, Acetic acid, phenyl- (6CI,8CI), 2-Phenylacetic acid, NSC 125718, PAA, Phenylethanoic acid, alpha-Toluic acid, omega-Phenylacetic acid, BRL-2853, Benzylpenicillin Potassium Ph Eur Impurity B, BRL-955, USP Amoxicillin Related Compound A, Ph Eur Amoxicillin Impurity A, Penicinic acid (1), Penicinic acid (2), 8-Hydroxypenillic acid (8-HPA), Phenylacetic acid, Tropicamide Related Compound D, Benzylpenicillin Benzathine Impurity B, Benzylpenicillin Potassium Impurity B, Benzylpenicillin Procaine Impurity E, Carbenicillin Sodium Impurity B, Cyclopentolate Hydrochloride Impurity B, Tropicamide Impurity D, Carbenicillin Impurity B, Cyclopentolate Impurity B, Benzylpenicillin Impurity B
InChI Formula InChI=1S/C8H8O2/c9-8(10)6-7-4-2-1-3-5-7/h1-5H,6H2,(H,9,10)
SMILES OC(=O)Cc1ccccc1
IUPAC Name 2-phenylacetic acid
Molecular Weight (g/mol) 136.15
Quantity 25 mg
Formula Weight 136.0524 g/mol
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.