missing translation for 'onlineSavingsMsg'
Learn More

Phenyl vinyl sulfone, 99+%

Product Code. p-7022161
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
This item is not returnable. View return policy

Product Code. 11452527

Brand: Thermo Scientific Alfa Aesar A14794.03

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 5535-48-8 | C8H8O2S | 168.21 g/mol

Phenyl vinyl sulfone is used as building blocks in the synthesis of a number of organic substances, participate in polymerization reactions, nucleophilic addition reaction, and in cycloaddition reactions. It is also used in the used in the synthesis of reactive dyes, synthesis of textile fibers, in particular cellulose fibers such as cotton and linen. Other applications in the field of medicine includes as powerful inhibitors of certain enzymatic processes. It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dye stuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Phenyl vinyl sulfone is used as building blocks in the synthesis of a number of organic substances, participate in polymerization reactions, nucleophilic addition reaction, and in cycloaddition reactions. It is also used in the used in the synthesis of reactive dyes, synthesis of textile fibers, in particular cellulose fibers such as cotton and linen. Other applications in the field of medicine includes as powerful inhibitors of certain enzymatic processes. It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dye stuff.

Solubility
It is soluble in most common organic solvents.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5535-48-8
Molecular Formula C8H8O2S
Molecular Weight (g/mol) 168.21
MDL Number MFCD00007554
InChI Key UJTPZISIAWDGFF-UHFFFAOYSA-N
Synonym phenyl vinyl sulfone, vinylsulfonyl benzene, ethenesulfonyl benzene, benzene, ethenylsulfonyl, sulfone, phenyl vinyl, vinylsulfonylbenzene, phenyl vinyl sulphone, ethenylsulfonyl benzene, ethenesulfonyl-benzene, phenyl-vinylsulfon
PubChem CID 79664
IUPAC Name ethenylsulfonylbenzene
SMILES C=CS(=O)(=O)C1=CC=CC=C1

Specifications

Melting Point 67°C to 71°C
Quantity 1 g
Beilstein 1906894
Solubility Information It is soluble in most common organic solvents.
Formula Weight 168.22
Percent Purity ≥99%
Chemical Name or Material Phenyl vinyl sulfone

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.