missing translation for 'onlineSavingsMsg'
Learn More

Phenyl vinyl sulfide, 97%, Thermo Scientific™

Product Code. 10521081 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 mL
Packaging:
Glass bottle
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10521081 1 mL Glass bottle 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10521081 Supplier Thermo Scientific Acros Supplier No. 210150010
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Specifications

CAS 1822-73-7
Density 1.0400g/mL
Boiling Point 94°C to 95°C (25.0 mmHg)
Flash Point 45°C
Assay Percent Range 97%
Packaging Glass bottle
Molecular Formula C8H8S
Linear Formula C6H5SCH=CH2
Refractive Index 1.5877 to 1.5897
MDL Number MFCD00009646
Quantity 1 mL
Beilstein 06, II, 288
Specific Gravity 1.04
Synonym phenyl vinyl sulfide, phenylthioethene, phenyl vinyl sulphide, phenylthio ethylene, vinylthiobenzene, benzene, ethenylthio, vinyl phenyl sulfide, sulfide, phenyl vinyl, vinylsulfanyl benzene, phenyl vinyl thioether
InChI Key GMPDOIGGGXSAPL-UHFFFAOYSA-N
SMILES C=CSC1=CC=CC=C1
IUPAC Name ethenylsulfanylbenzene
Molecular Weight (g/mol) 136.21
PubChem CID 74572
Formula Weight 136.21
Percent Purity 97%
Chemical Name or Material Phenyl vinyl sulfide, 97%
Show More Show Less
compliance-icons
Product Identifier
  • Phenyl vinyl sulfide
Signal Word
  • Warning
Hazard Category
  • Flammable liquid Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.