missing translation for 'onlineSavingsMsg'
Learn More

Phenyl trifluoroacetate, 98%

Product Code. 11429217
Change view
Click to view available options
Quantity:
25 g
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11429217 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11429217 Supplier Thermo Scientific Alfa Aesar Supplier No. B21304.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 500-73-2 | C8H5F3O2 | 190.121 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 500-73-2
Molecular Formula C8H5F3O2
Molecular Weight (g/mol) 190.121
MDL Number MFCD00039232
InChI Key DVCMYAIUSOSIQP-UHFFFAOYSA-N
Synonym phenyl trifluoroacetate, acetic acid, trifluoro-, phenyl ester, acetic acid, 2,2,2-trifluoro-, phenyl ester, trifluoroacetic acid phenyl ester, phenyltrifluoroacetate, acmc-209kjz, dvcmyaiusosiqp-uhfffaoysa
PubChem CID 68143
IUPAC Name phenyl 2,2,2-trifluoroacetate
SMILES C1=CC=C(C=C1)OC(=O)C(F)(F)F

Specifications

Density 1.276
Boiling Point 146°C to 147°C
Flash Point 52°C (127°F)
Refractive Index 1.423
Quantity 25 g
UN Number UN3272
Sensitivity Moisture sensitive
Formula Weight 190.12
Percent Purity 98%
Chemical Name or Material Phenyl trifluoroacetate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.