missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Phenyl Sepharose™ High Performance Media

Product Code. 10668502 Shop All Cytiva Products
Change view
Click to view available options
Quantity:
75 mL
Unit Size:
Each
Product Code. Quantity unitSize
10668502 75 mL Each
1 options
This item is not returnable. View return policy

Product Code. 10668502

Brand: Cytiva 17108201

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Designed for intermediate and polishing step purification steps

  • Hydrophobic interaction chromatography (HIC) media composed of highly cross-linked agarose beads with aromatic phenyl functional groups
  • High resolution due to smaller 34μm bead size
  • Chemical stability for effective CIP and sanitization
  • Available in prepacked columns such as HiPrep™, HiScreen™, and HiTrap™ plus laboratory and BioProcess pack sizes

TRUSTED_SUSTAINABILITY

Specifications

Quantity 75 mL
Stability Stable in commonly used aqueous Buffers: 1 M NaOH, 1 M Acetic Acid, 30% Acetonitrile, 70% Ethanol, 30% Isopropanol, 6 M Guanidine-Hydrochloride, 8 M Urea, , 3 M Ammonium sulfate, Ionic Detergents, Non-ionic detergents, polar organic solvents, 2% Sodium Dodectyl Sulfate
For Use With (Application) For Intermediate PurifiCatIon and Polishing AppliCatIons
Storage Requirements 4°C to 30°C
Product Type Phenyl Sepharose High Performance Media
Flow Rate 100 to 200 cm/Hoμr
Particle Size 24 to 44 μm
Height (English) Bed 11.8 in.
Height (Metric) Bed 30 cm
Max. Pressure 300 kPa
Matrix 6% Cross-Linked Agarose
pH Range 3 to 13
Storage Buffer 20% Ethanol
Ligand Type Phenyl
Show More Show Less
compliance-icons
Product Identifier
  • Phenyl Sepharose High Performance
Signal Word
  • Warning
Hazard Category
  • Flammable liquid Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contains : ethanol
  • UNKNOWN ACUTE-16.5 per cent of the mixture consists of component(s) of unknown acute toxicity.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.