missing translation for 'onlineSavingsMsg'
Learn More

Phenyl selenocyanate, 98%

Product Code. 11359026
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11359026 1 g 1g
11369026 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11359026 Supplier Thermo Scientific Alfa Aesar Supplier No. L20194.03
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 2179-79-5 | C7H5NSe | 182.095 g/mol

Phenyl selenocyanate is used as a specific source of nucleophilic and electrophilic selenium species in synthetic chemistry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Phenyl selenocyanate is used as a specific source of nucleophilic and electrophilic selenium species in synthetic chemistry.

Solubility
Miscible with organic solvents, terahydro furan, dichloromethane and acetonitrile.

Notes
Store in a cool place. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2179-79-5
Molecular Formula C7H5NSe
Molecular Weight (g/mol) 182.095
MDL Number MFCD00216944
InChI Key NODWRXQVQYOJGN-UHFFFAOYSA-N
Synonym phenylselenocyanate, phenylselanyl formonitrile, selenocyanic acid, phenyl ester, acmc-1cgab, phenyl selenocyanate, selenocyanic acid phenyl ester, 2-phenyl-2-selenaethanenitrile
PubChem CID 555340
IUPAC Name phenyl selenocyanate
SMILES C1=CC=C(C=C1)[Se]C#N

Specifications

Density 1.484
Boiling Point 116°C to 117°C (12 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.603
Quantity 1 g
UN Number UN3440
Beilstein 1617370
Solubility Information Miscible with organic solvents,terahydro furan,dichloromethane and acetonitrile.
Formula Weight 182.08
Percent Purity 98%
Chemical Name or Material Phenyl selenocyanate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.