missing translation for 'onlineSavingsMsg'
Learn More

Phenyl Salicylate, Mikromol™

Product Code. 17900756
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17900756 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17900756 Supplier Mikromol Supplier No. MM1884.00
 Restricted Item
Estimated Shipment: 23-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Phenyl Salicylate
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Phenyl Salicylate
CAS 118-55-8
Analyte or Component Names Phenyl Salicylate
Shipping Condition Room Temperature
Molecular Formula C13 H10 O3
Certifications/Compliance ISO 17034
Synonym Benzoic acid, 2-hydroxy-, phenyl ester, Salicylic acid, phenyl ester (8CI), 2-Phenoxycarbonylphenol, NSC 33406, Phenol salicylate, Phenyl 2-hydroxybenzoate, Phenyl salicylate, Salol, Salphenyl, Seesorb 201, Seesorb K 201, Phenyl Salicylate
InChI Formula InChI=1S/C13H10O3/c14-12-9-5-4-8-11(12)13(15)16-10-6-2-1-3-7-10/h1-9,14H
SMILES Oc1ccccc1C(=O)Oc2ccccc2
IUPAC Name phenyl 2-hydroxybenzoate
Molecular Weight (g/mol) 214.22
Quantity 250 mg
Formula Weight 214.063 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.