missing translation for 'onlineSavingsMsg'
Learn More

Phenyl phosphorodichloridate, 97%, Thermo Scientific Chemicals

Catalog Number 11408723
Click to view available options
Quantity:
25 g
100 g
500 g
This item is not returnable. View return policy
770-12-7
C6H5Cl2O2P
210.978
MFCD00002067
TXFOLHZMICYNRM-UHFFFAOYSA-N
phenyl dichlorophosphate, phenyl phosphorodichloridate, dichlorophenoxyphosphine oxide, phosphorodichloridic acid, phenyl ester, phenylphosphoric dichloride, phenyl phosphorodichlorodate, phenoxydichlorophosphine oxide, phenyldichlorophosphate, phenoxyphosphoryl dichloride, phosphoroyl dichloride phenyl ester
13038
dichlorophosphoryloxybenzene
C1=CC=C(C=C1)OP(=O)(Cl)Cl
This item is not returnable. View return policy
770-12-7
C6H5Cl2O2P
210.978
MFCD00002067
TXFOLHZMICYNRM-UHFFFAOYSA-N
phenyl dichlorophosphate, phenyl phosphorodichloridate, dichlorophenoxyphosphine oxide, phosphorodichloridic acid, phenyl ester, phenylphosphoric dichloride, phenyl phosphorodichlorodate, phenoxydichlorophosphine oxide, phenyldichlorophosphate, phenoxyphosphoryl dichloride, phosphoroyl dichloride phenyl ester
13038
dichlorophosphoryloxybenzene
C1=CC=C(C=C1)OP(=O)(Cl)Cl

Reagent for preparation of phosphate diesters.Phenyl phosphorodichloridate is used in the preparation of symmetrical phosphate diesters. It is used as a phosphorylating agent for alcohols and amines. It is also involved in the preparation of phenylphosphoro di-(1-imidazolidate) and 2-phenyl-bis-triazoloylphosphate, which is used in peptide synthesis. It plays a vital role in the Pfitzner-Moffatt oxidation which involves oxidation of alcohols to the corresponding aldehydes and ketones. Furthermore, it is utilized in Beckmann rearrangement for the preparation of N-phenylacetamide from acetophenone oxime.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Reagent for preparation of phosphate diesters.Phenyl phosphorodichloridate is used in the preparation of symmetrical phosphate diesters. It is used as a phosphorylating agent for alcohols and amines. It is also involved in the preparation of phenylphosphoro di-(1-imidazolidate) and 2-phenyl-bis-triazoloylphosphate, which is used in peptide synthesis. It plays a vital role in the Pfitzner-Moffatt oxidation which involves oxidation of alcohols to the corresponding aldehydes and ketones. Furthermore, it is utilized in Beckmann rearrangement for the preparation of N-phenylacetamide from acetophenone oxime.

Solubility
Immiscible with water.

Notes
Moisture sensitive. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with strong bases, alcohols and strong oxidizing agents.
TRUSTED_SUSTAINABILITY
Melting Point -1°C
Density 1.415
Boiling Point 241°C to 243°C
Flash Point 112°C (233°F)
Refractive Index 1.523
Quantity 25 g
UN Number UN3265
Beilstein 511863
Sensitivity Moisture sensitive
Solubility Information Immiscible with water.
Formula Weight 210.98
Percent Purity 97%
Chemical Name or Material Phenyl phosphorodichloridate
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'