missing translation for 'onlineSavingsMsg'
Learn More

Phenyl phosphate disodium hydrate, TRC

Product Code. 30410547
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30410547 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30410547 Supplier Toronto Research Chemicals Supplier No. TRCP3187005G
 Restricted Item
Estimated Shipment: 13-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3279-54-7
Molecular Formula C6H5Na2O4P . 2(H2O)
Molecular Weight (g/mol) 218.05
Synonym Disodiumphenylphosphate,dihyy, Phosphoric acid, monophenylester, disodium salt, Disodium phenyl-(o)-phosphate, Di-sodium phenyl phosphate dihydrate, Disodium phenylphosphate, Phenylphosphoric acid disodium salt, Phenyl phosphate disodium salt, Phenyl disodium phosphate
IUPAC Name disodium;phenyl phosphate
SMILES [Na+].[Na+].[O-]P(=O)([O-])Oc1ccccc1

Specifications

Quantity 5 g
InChI Formula InChI=1S/C6H7O4P.2Na/c7-11(8,9)10-6-4-2-1-3-5-6;;/h1-5H,(H2,7,8,9);;/q;2*+1/p-2
Formula Weight 217.9721
Chemical Name or Material Phenyl phosphate disodium hydrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.