missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phenyl N-iso-Propyl-d7-carbamate, CDN
Click to view available options
Quantity:
0.1 g
0.25 g
Unit Size:
0.1g
0.25g
Specifications
Specifications
| Chemical Name or Material | (1-Methylethyl)carbamic Acid-d7 Phenyl Ester |
| CAS | 1329835-22-4 |
| Molecular Formula | C10 D7 H6 N O2 |
| Quantity | 0.25 g |
| Synonym | Phenyl N-(1,1,1,2,3,3,3-heptadeuteriopropan-2-yl)carbamate, Isopropyl-d7-carbamic acid phenyl ester, Phenyl N-[1-(methyl-d3)ethyl-1,2,2,2-d4]carbamate, Phenyl N-(isopropyl-d7)carbamate, [1-(Methyl-d3)ethyl-1,2,2,2-d4]carbamic acid phenyl ester, Phenyl isopropylcarbamate-D7 (isopropyl-D7) |
| InChI Formula | InChI=1 S/C10H13NO2/c1-8(2)11-10(12)13-9-6-4-3-5-7-9/h3-8 H,1-2H3,(H,11,12)/i1D3,2D3,8 D |
| SMILES | [2 H]C([2 H])([2 H])C([2 H])(NC(=O)Oc1ccccc1)C([2 H])([2 H])[2 H] |
| IUPAC Name | phenyl N-(1,1,1,2,3,3,3-heptadeuteriopropan-2-yl)carbamate |
| Molecular Weight (g/mol) | 186.259 |
| Formula Weight | 186.139 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction