missing translation for 'onlineSavingsMsg'
Learn More

Phenyl isothiocyanate, 98%

Product Code. 10245780 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Glass bottle
Unit Size:
100g
2.5kg
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10245780 500 g Glass bottle 500g
10503911 100 g Glass bottle 100g
11925731 2.5 kg Glass bottle 2.5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10245780 Supplier Thermo Scientific Acros Supplier No. 160975000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 103-72-0 | C7H5NS | 135.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 103-72-0
Molecular Formula C7H5NS
Molecular Weight (g/mol) 135.18
MDL Number MFCD00004798
InChI Key QKFJKGMPGYROCL-UHFFFAOYSA-N
Synonym phenyl isothiocyanate, phenylisothiocyanate, thiocarbanil, phenyl mustard oil, benzene, isothiocyanato, pitc, isothiocyanic acid phenyl ester, benzene-1-isothiocyanate, phenylsenfoel, phenyl thioisocyanate
PubChem CID 7673
ChEBI CHEBI:85103
IUPAC Name isothiocyanatobenzene
SMILES S=C=NC1=CC=CC=C1

Specifications

Melting Point -21.0°C
Color Yellow
Density 1.1288g/mL
Boiling Point 221.0°C
Flash Point 87°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula C6H5NCS
Refractive Index 1.6490 to 1.6510
Quantity 500 g
Beilstein 12,453
Fieser 01,844; 02,323
Merck Index 15,7408
Specific Gravity 1.1288
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in ethanol and diethyl ether
Viscosity 1.3 mPa.s (20°C)
Formula Weight 135.19
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material Phenyl isothiocyanate
Show More Show Less
compliance-icons
Hazard Category
  • Acute toxicity Category 3
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Serious eye damage/eye irritation Category 1
  • Respiratory sensitiser Category 1
  • Skin corrosion/irritation Category 1 B
  • Skin sensitiser Category 1
Hazard Statement
  • H301-Toxic if swallowed.
  • H314-Causes severe skin burns and eye damage.
  • H317-May cause an allergic skin reaction.
  • H318-Causes serious eye damage.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P284-¢In case of inadequate ventilation! wear respiratory protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.