missing translation for 'onlineSavingsMsg'
Learn More

Phenyl isothiocyanate, 97%

Product Code. 11464673
Change view
Click to view available options
Quantity:
5 g
100 g
500 g
Unit Size:
100g
500g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11464673 500 g 500g
11414040 5 g 5g
11454673 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11464673 Supplier Thermo Scientific Alfa Aesar Supplier No. A11596.36
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 103-72-0 | C7H5NS | 135.18 g/mol

Phenyl isothiocyanate acts as a derivatizing reagent for primary and secondary amines. It is used in sequencing peptides by Edman degradation and in amino acid analyses by HPLC. It is used for derivatizing N-terminal amino acids of proteins for automated sequential analysis. It is a synthon for dithiadiazafulvalenes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Phenyl isothiocyanate acts as a derivatizing reagent for primary and secondary amines. It is used in sequencing peptides by Edman degradation and in amino acid analyses by HPLC. It is used for derivatizing N-terminal amino acids of
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 103-72-0
Molecular Formula C7H5NS
Molecular Weight (g/mol) 135.18
MDL Number MFCD00004798
InChI Key QKFJKGMPGYROCL-UHFFFAOYSA-N
Synonym phenyl isothiocyanate, phenylisothiocyanate, thiocarbanil, phenyl mustard oil, benzene, isothiocyanato, pitc, isothiocyanic acid phenyl ester, benzene-1-isothiocyanate, phenylsenfoel, phenyl thioisocyanate
PubChem CID 7673
ChEBI CHEBI:85103
IUPAC Name isothiocyanatobenzene
SMILES S=C=NC1=CC=CC=C1

Specifications

Melting Point -21°C
Density 1.131
Boiling Point 220°C to 221°C
Flash Point 87°C (188°F)
Odor Pungent
Refractive Index 1.65
Quantity 500 g
UN Number UN2927
Beilstein 471392
Sensitivity Moisture sensitive
Merck Index 14,7297
Solubility Information Soluble in alcohol,and ether. Insoluble in water.
Formula Weight 135.19
Percent Purity 97%
Chemical Name or Material Phenyl isothiocyanate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.