missing translation for 'onlineSavingsMsg'
Learn More

Phenyl isocyanate, 99%

Product Code. 10225230 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
100 g
250 g
1 kg
Packaging:
Glass bottle
Unit Size:
100g
1kg
250g
5g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10225230 100 g Glass bottle 100g
10215230 5 g Glass bottle 5g
10143420 250 g Glass bottle 250g
10795591 1 kg Glass bottle 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 10225230 Supplier Thermo Scientific Acros Supplier No. 130691000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 103-71-9 | C7H5NO | 119.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 103-71-9
MDL Number MFCD00001994
InChI Key DGTNSSLYPYDJGL-UHFFFAOYSA-N
Synonym phenyl isocyanate, phenylcarbimide, carbanil, phenyl carbonimide, benzene, isocyanato, phenylisocyanate, isocyanic acid, phenyl ester, mondur p, phnco, fenylisokyanat
PubChem CID 7672
ChEBI CHEBI:53806
IUPAC Name isocyanatobenzene
SMILES C1=CC=C(C=C1)N=C=O

Specifications

Color Colorless to Yellow
Density 1.095g/mL
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Molecular Formula C7H5NO
Linear Formula C6H5NCO
Refractive Index 1.5350 to 1.5370 (20°C, 589nm)
Quantity 100 g
Beilstein 12, 437
Fieser 01,842; 02,322; 04,378; 07,284; 08,396; 10,309; 12,386
Merck Index 15, 7407
Molecular Weight (g/mol) 119.123
Formula Weight 119.12
Percent Purity ≥98.5%
Physical Form Liquid
Chemical Name or Material Phenyl isocyanate, 99%
Show More Show Less
compliance-icons compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • Phenyl isocyanate
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 3
  • Acute toxicity Category 4
  • Acute toxicity Category 1
  • Skin corrosion/irritation Category 1 C
  • Serious eye damage/eye irritation Category 1
  • Respiratory sensitiser Category 1
  • Skin sensitiser Category 1
  • Specific target organ toxicity Category 3
  • ACUTE AQUATIC Acute 1
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
  • H317-May cause an allergic skin reaction.
  • H330-Fatal if inhaled.
  • H302-Harmful if swallowed.
  • H314-Causes severe skin burns and eye damage.
  • H335-May cause respiratory irritation.
  • H400-Very toxic to aquatic life.
  • H411-Toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.