missing translation for 'onlineSavingsMsg'
Learn More

Phenyl formate, 95%

Product Code. p-7043333
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11369973 5 g 5g
11379973 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11369973 Supplier Thermo Scientific Alfa Aesar Supplier No. L00289.06
Add to basket
This item is not returnable. View return policy

CAS: 1864-94-4 | C7H6O2 | 122.123 g/mol

Phenyl formate is widely used in the palladium-catalyzed carbonylation of aryl, alkenyl and allyl halides. It is also used as a reagent for the formylation of amines.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Phenyl formate is widely used in the palladium-catalyzed carbonylation of aryl, alkenyl and allyl halides. It is also used as a reagent for the formylation of amines.

Solubility
Immiscible with water, alcohol, ether and benzene.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1864-94-4
Molecular Formula C7H6O2
Molecular Weight (g/mol) 122.123
MDL Number MFCD00014126
InChI Key GEOWCLRLLWTHDN-UHFFFAOYSA-N
Synonym formic acid phenyl ester, formic acid, phenyl ester, phocho, formic acid, phenylester, acmc-1box2, phenyl formate gc
PubChem CID 74626
IUPAC Name phenyl formate
SMILES C1=CC=C(C=C1)OC=O

Specifications

Density 1.09
Boiling Point 72°C to 74°C (13 mmHg)
Flash Point 71°C (159°F)
Refractive Index 1.512
Quantity 5 g
Beilstein 507113
Solubility Information Immiscible with water,alcohol,ether and benzene.
Formula Weight 122.12
Percent Purity 95%
Chemical Name or Material Phenyl formate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.