missing translation for 'onlineSavingsMsg'
Learn More

Phenyl Ether-biphenyl Eutectic, Thermo Scientific Chemicals

Catalog Number p-182794
Click to view available options
Quantity:
100 g
1 kg
This item is not returnable. View return policy
8004-13-5
C24H20O
324.41
MFCD00148859
MHCVCKDNQYMGEX-UHFFFAOYSA-N
diphyl, dowtherm, dowtherm a, dinil, dinyl, therminol vp, phenyl ether-biphenyl mixture, phenyl ether-diphenyl mixture, biphenyl-diphenyl ether mixture, hsdb 137
24670
1,1'-biphenyl;phenoxybenzene
C1=CC=C(C=C1)C2=CC=CC=C2.C1=CC=C(C=C1)OC2=CC=CC=C2
This item is not returnable. View return policy
8004-13-5
C24H20O
324.41
MFCD00148859
MHCVCKDNQYMGEX-UHFFFAOYSA-N
diphyl, dowtherm, dowtherm a, dinil, dinyl, therminol vp, phenyl ether-biphenyl mixture, phenyl ether-diphenyl mixture, biphenyl-diphenyl ether mixture, hsdb 137
24670
1,1'-biphenyl;phenoxybenzene
C1=CC=C(C=C1)C2=CC=CC=C2.C1=CC=C(C=C1)OC2=CC=CC=C2

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 13°C
Density 1.0560g/mL
Flash Point 123°C
Packaging Glass bottle
Refractive Index 1.5885 to 1.5905
Quantity 100 g
Specific Gravity 1.056
Formula Weight 324.41
Chemical Name or Material Phenyl ether-biphenyl eutectic
compliance-icons compliance-icons
Product Identifier
  • Phenyl ether-biphenyl eutectic
Signal Word
  • Warning
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'