missing translation for 'onlineSavingsMsg'
Learn More

Phenyl-d5-acetic Acid, CDN

Product Code. 30322114
Change view
Click to view available options
Quantity:
0.5 g
1 g
Unit Size:
0.5g
1g
Product Code. Quantity unitSize
30322114 1 g 1g
30319533 0.5 g 0.5g
2 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Product Code. 30322114

Brand: CDN CDND28201G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Phenylacetic Acid D5 (phenyl-D5)
CAS 104182-98-1
Molecular Formula C8 D5 H3 O2
Quantity 1 g
Synonym Benzene-2,3,4,5,6-d5-acetic acid (ACI), Benzene-d5-acetic acid (9 CI), Phenyl-d5-acetic acid, Phenylacetic acid-d5, Phenylacetic acid-d5
InChI Formula InChI=1 S/C8H8O2/c9-8(10)6-7-4-2-1-3-5-7/h1-5 H,6H2,(H,9,10)/i1D,2 D,3 D,4 D,5 D
SMILES [2 H]c1c([2 H])c([2 H])c(CC(=O)O)c([2 H])c1[2 H]
IUPAC Name 2-(2,3,4,5,6-pentadeuteriophenyl)acetic acid
Molecular Weight (g/mol) 141.1787
Formula Weight 141.0838 g/mol
Percent Purity 98 atom % D, min 98% Chemical Purity
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.